CAS RN 6061-96-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-Methylcitric Acid, TRC
CAS: 6061-96-7 Molecular Formula: C7 H10 O7 Molecular Weight (g/mol): 206.15 Synonym: 1,2,3-Butanetricarboxylic acid, 2-hydroxy- (7CI, 8CI, 9CI, ACI),2-Hydroxy-1,2,3-butanetricarboxylic acid (ACI),2-Hydroxy-1-methylpropane-1,2,3-tricarboxylic acid,2-Methylcitric acid IUPAC Name: 2-hydroxybutane-1,2,3-tricarboxylic acid SMILES: CC(C(=O)O)C(O)(CC(=O)O)C(=O)O
2
2-Methylcitric Acid (mixture of diastereomers), CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.