Learn More
2-Methylbenzimidazole, 98%
Product Code. 11493517
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493517 1 kg 1000g
11403527 50 g 50g
11413527 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11493517 Supplier Thermo Scientific Alfa Aesar Supplier No. A17290.0B
 Restricted Item
In Stock
Add to basket
CAS: 615-15-6 | C8H8N2 | 132.17 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 615-15-6
Molecular Formula C8H8N2
Molecular Weight (g/mol) 132.17
MDL Number MFCD00005598
InChI Key LDZYRENCLPUXAX-UHFFFAOYSA-N
PubChem CID 11984
SMILES CC1=NC2=CC=CC=C2N1
Specifications
Melting Point 174°C to 178°C
Quantity 1 kg
Beilstein 112264
Synonym 2-methylbenzimidazole, 2-methyl-1 h-benzo d imidazole, 1 h-benzimidazole, 2-methyl, benzimidazole, 2-methyl, 2-methyl-1 h-benzoimidazole, 2-methyl-1h-1,3-benzodiazole, acetamidine, n-n'-o-phenylene, 2-methyl benzimidazole, unii-zh8iww7y8b, 2-methylbenaimidazole
IUPAC Name 2-methyl-1H-1,3-benzodiazole
Formula Weight 132.17
Percent Purity 98%
Chemical Name or Material 2-Methylbenzimidazole