accessibility menu, dialog, popup

UserName

H3CHNN

CAS RN 615-15-6

IUPAC Name: 2-methyl-1H-1,3-benzodiazole
Synonyms: 2-methylbenaimidazole
Molecular Weight (g/mol): 132.17
Molecular Formula: C8H8N2
InChi Key: LDZYRENCLPUXAX-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=CC=C2N1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (5)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)