Learn More
2-Methyl-5-(trifluoromethyl)aniline, 98%, Thermo Scientific™
Product Code. 11415566
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415566 1 g 1g
11425566 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11415566 Supplier Thermo Scientific Alfa Aesar Supplier No. B20709.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25449-96-1
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.154
MDL Number MFCD00134580
InChI Key BCLCKENDTZITFB-UHFFFAOYSA-N
Synonym 3-amino-4-methylbenzotrifluoride, 2-methyl-5-trifluoromethyl aniline, benzenamine, 2-methyl-5-trifluoromethyl, 2-methyl-5-trifluoromethyl phenylamine, pubchem4494, intermediates-zcf02681, acmc-209gk1, 5-trifluoromethyl-o-toluidine, 5-trifluoromethyl-2-methylaniline
PubChem CID 2735933
IUPAC Name 2-methyl-5-(trifluoromethyl)aniline
SMILES CC1=C(C=C(C=C1)C(F)(F)F)N
Specifications
Melting Point 36°C to 40°C
Quantity 1 g
UN Number UN2811
Formula Weight 175.16
Percent Purity 98%
Chemical Name or Material 2-Methyl-5-(trifluoromethyl)aniline