accessibility menu, dialog, popup

UserName

CAS RN 25449-96-1

IUPAC Name: 2-methyl-5-(trifluoromethyl)aniline
Synonyms: 2-methyl-5-(trifluoromethyl)aniline
Molecular Weight (g/mol): 175.15
Molecular Formula: C8H8F3N
InChi Key: BCLCKENDTZITFB-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1N)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results