Learn More
2-Methyl-4-phenyl-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™
Product Code. p-4510653
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11320617 250 mg Amber Glass Bottle 250mg
10071884 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11320617 Supplier Thermo Scientific Maybridge Supplier No. CC42201CB
Chemical Identifiers
CAS 32002-72-5
Molecular Formula C11H8NO2S
Molecular Weight (g/mol) 218.25
MDL Number MFCD07348569
InChI Key APXSARIYDLVPAY-UHFFFAOYSA-M
Synonym 2-methyl-4-phenylthiazole-5-carboxylic acid, 2-methy-4-phenylthiazol-5-ylcarboxylic acid, 2-methyl-4-phenyl-thiazole-5-carboxylic acid, 2-methyl-4-phenylthiazol-5-ylcarboxylic acid, 5-thiazolecarboxylicacid, 2-methyl-4-phenyl, 5-thiazolecarboxylic acid, 2-methyl-4-phenyl
PubChem CID 7131132
IUPAC Name 2-methyl-4-phenyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=NC(=C(S1)C([O-])=O)C1=CC=CC=C1
Specifications
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 219.26
Percent Purity 97%
Chemical Name or Material 2-Methyl-4-phenyl-1, 3-thiazole-5-carboxylic acid