accessibility menu, dialog, popup

UserName

CAS RN 32002-72-5

CH3SOON

CAS RN 32002-72-5

IUPAC Name: 2-methyl-4-phenyl-1,3-thiazole-5-carboxylate
Synonyms: 2-methyl-4-phenyl-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 218.25
Molecular Formula: C11H8NO2S
InChi Key: APXSARIYDLVPAY-UHFFFAOYSA-M
SMILES: CC1=NC(=C(S1)C([O-])=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results