missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypropanol, TRC

Product Code. 30007359
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007359 10 g 10g
17979859 1 g 1g
17999859 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007359 Supplier Toronto Research Chemicals Supplier No. TRCM22725510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1589-47-5
Molecular Formula C4 H10 O2
Molecular Weight (g/mol) 90.12
Synonym 2-Methoxy-1-propanol, 2-Methoxy-1-hydroxypropane, 2-Methoxypropanol
IUPAC Name 2-methoxypropan-1-ol
SMILES COC(C)CO

Specifications

Quantity 10 g
InChI Formula InChI=1S/C4H10O2/c1-4(3-5)6-2/h4-5H,3H2,1-2H3
Formula Weight 90.0681
Percent Purity >95%
Chemical Name or Material 2-Methoxypropanol
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.