CAS RN 1589-47-5
CAS RN 1589-47-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Methoxypropanol, TRC
CAS: 1589-47-5 Molecular Formula: C4 H10 O2 Molecular Weight (g/mol): 90.12 Synonym: 2-Methoxy-1-propanol,2-Methoxy-1-hydroxypropane,2-Methoxypropanol IUPAC Name: 2-methoxypropan-1-ol SMILES: COC(C)CO