Learn More
2-Keto-D-gluconic acid, hemicalcium salt monohydrate, >98%, MP Biomedicals™
Product Code. 11453050 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11453050 1 g 1g
11463050 5 g 5g
11473050 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11453050 Supplier MP Biomedicals Supplier No. 0210036501
  • ONLINE EXCLUSIVE PRICE
    261.00 EUR
    247.80 EUR / 1g
    Save 13.20 EUR
    5% Off
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
2-Keto-D-gluconic Acid hemicalcium salt is Hemicalcium Salt of 2-Keto-D-gluconic acid. It is produced from D-glucose
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3470-37-9
Molecular Formula C12H20CaO14
Molecular Weight (g/mol) 428.356
MDL Number MFCD00149104
InChI Key QACANIDDQFGAJU-JQTJLMCZSA-N
Synonym calcium 2-keto-d-gluconate
PubChem CID 131852723
IUPAC Name calcium;(3S,4R,5R)-3,4,5,6-tetrahydroxy-2-oxohexanoic acid
SMILES C(C(C(C(C(=O)C(=O)O)O)O)O)O.C(C(C(C(C(=O)C(=O)O)O)O)O)O.[Ca]
Specifications
Color White
Assay Percent Range >98%
Quantity 1 g
Formula Weight 213.2
Physical Form Powder
Chemical Name or Material 2-Keto-D-gluconic acid, hemicalcium salt