accessibility menu, dialog, popup

UserName

OOOHOHOHOHHOOOOHOHOHOHHOCa

CAS RN 3470-37-9

IUPAC Name: bis(3,4,5,6-tetrahydroxy-2-oxohexanoic acid) calcium
Synonyms: bis(2-keto-D-gluconic acid) calcium
Molecular Weight (g/mol): 428.36
Molecular Formula: C12H20CaO14
InChi Key: QACANIDDQFGAJU-UHFFFAOYNA-N
SMILES: [Ca].OCC(O)C(O)C(O)C(=O)C(O)=O.OCC(O)C(O)C(O)C(=O)C(O)=O
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results