missing translation for 'onlineSavingsMsg'
Learn More

2-Iminothiolane, MP Biomedicals™

Product Code. 11457981 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11457981 100 mg 100mg
11467981 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11457981 Supplier MP Biomedicals Supplier No. 0215131980
  • ONLINE EXCLUSIVE PRICE
    109.00 EUR
    102.90 EUR / 100mg
    Save 6.10 EUR
    5% Off
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 4781-83-3
Molecular Formula C4H8ClNS
Molecular Weight (g/mol) 137.625
InChI Key ATGUDZODTABURZ-UHFFFAOYSA-N
Synonym 2-iminothiolane hydrochloride, 2-iminothiolane hcl, thiolan-2-imine hydrochloride, dihydrothiophen-2 3h-imine hydrochloride, 2 3h-thiophenimine, dihydro-, hydrochloride, traut's reagent, c4h7ns.clh, traut eo 1/4 a, 2-thiolanimine hydrochloride, 2 3h-thiophenimine, hydrochloride
PubChem CID 13166855
IUPAC Name thiolan-2-imine;hydrochloride
SMILES C1CC(=N)SC1.Cl

Specifications

Color White
Assay Percent Range 0.98
Quantity 100 mg
Formula Weight 137.3
Physical Form Crystalline Powder
Chemical Name or Material 2-Iminothiolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.