accessibility menu, dialog, popup

UserName

CAS RN 4781-83-3

HClNHS

CAS RN 4781-83-3

IUPAC Name: hydrogen thiolan-2-imine chloride
Synonyms: 2-iminothiolane
Molecular Weight (g/mol): 137.63
Molecular Formula: C4H8ClNS
InChi Key: ATGUDZODTABURZ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].N=C1CCCS1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)