missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethylammonium Chloride, Sigma-Aldrich™

Product Code. 15840731
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15840731 500 g 500g
15830731 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15840731 Supplier Sigma-Aldrich Supplier No. 8200630500
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

2-Hydroxyethylammonium chloride also known as ethanolammonium chloride used in the synthesis of esters and ethers via nucleophilic substitution reactions. It can be used to functionalization of various polymers.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2002-24-6
Grade Research
Molecular Weight (g/mol) 61.08 (Free Base Basis)

Specifications

Melting Point 80°C to 84°C
MDL Number MFCD00012892
Quantity 500 g
Synonym 2-Hydroxyethylammonium chloride, Ethanolamine hydrochloride, 2-Aminoethanol hydrochloride, Ethanolammonium chloride, 2-Hydroxyethylammonium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.