accessibility menu, dialog, popup

UserName

HClNH2HO

CAS RN 2002-24-6

IUPAC Name: hydrogen 2-aminoethan-1-ol chloride
Synonyms: ethanolamine hcl
Molecular Weight (g/mol): 97.54
Molecular Formula: C2H8ClNO
InChi Key: PMUNIMVZCACZBB-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCO
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)