missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy Hippuric Acid-13C2,15N, TRC

Product Code. 30180084
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180084 10 mg 10mg
30179874 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180084 Supplier Toronto Research Chemicals Supplier No. TRCH94288710MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1286521-95-6
Molecular Formula C713C2H915NO4
Molecular Weight (g/mol) 198.15
Synonym N-(2-Hydroxybenzoyl)glycine-13C2,15N, o-Hydroxy-hippuric Acid, Salicyluric Acid-13C2,15N, (2-Hydroxybenzoyl)glycine-13C2,15N, 2-Hydroxyhippuric Acid-13C2,15N, NSC 524135-13C2,15N, Salicyloylglycine-13C2,15N, o-Hydroxyhippuric Acid-13C2,15N;
IUPAC Name 2-[(2-hydroxybenzoyl)amino]acetic acid
SMILES OC1=CC=CC=C1C([15NH][13CH2][13C](O)=O)=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H9NO4/c11-7-4-2-1-3-6(7)9(14)10-5-8(12)13/h1-4,11H,5H2,(H,10,14)(H,12,13)/i5+1,8+1,10+1
Formula Weight 198.0569
Chemical Name or Material 2-Hydroxy Hippuric Acid-13C2,15N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.