CAS RN 1286521-95-6
CAS RN 1286521-95-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Hydroxy Hippuric Acid-13C2,15N, TRC
CAS: 1286521-95-6 Molecular Formula: C713C2H915NO4 Molecular Weight (g/mol): 198.15 Synonym: N-(2-Hydroxybenzoyl)glycine-13C2,15N,o-Hydroxy-hippuric Acid,Salicyluric Acid-13C2,15N,(2-Hydroxybenzoyl)glycine-13C2,15N,2-Hydroxyhippuric Acid-13C2,15N,NSC 524135-13C2,15N,Salicyloylglycine-13C2,15N,o-Hydroxyhippuric Acid-13C2,15N; IUPAC Name: 2-[(2-hydroxybenzoyl)amino]acetic acid SMILES: OC1=CC=CC=C1C([15NH][13CH2][13C](O)=O)=O