missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-4-methylbenzonitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Fluoro-4-methylbenzonitrile is used in the synthesis of fluorobenzamidrazone thrombin inhibitors.
Solubility
Insoluble in water, soluble in chloroform, acetone and other organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
2-Fluoro-4-methylbenzonitrile is used in the synthesis of fluorobenzamidrazone thrombin inhibitors.
Solubility
Insoluble in water, soluble in chloroform, acetone and other organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 52°C to 53°C |
| Quantity | 5 g |
| UN Number | UN3439 |
| Solubility Information | Insoluble in water,soluble in chloroform,acetone and other organic solvents. |
| Formula Weight | 135.14 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Fluoro-4-methylbenzonitrile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction