accessibility menu, dialog, popup

UserName

CAS RN 85070-67-3

CH3NF

CAS RN 85070-67-3

IUPAC Name: 2-fluoro-4-methylbenzonitrile
Synonyms: 2-fluoro-4-methylbenzonitrile
Molecular Weight (g/mol): 135.14
Molecular Formula: C8H6FN
InChi Key: WCGNLBCJPBKXCN-UHFFFAOYSA-N
SMILES: CC1=CC=C(C#N)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results