Learn More
2-(Diphenylphosphino)ethylamine, 95%
Product Code. p-7029592
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436130 1 g 1g
11446130 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11436130 Supplier Thermo Scientific Alfa Aesar Supplier No. H27070.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 4848-43-5 | C14H16NP | 229.26 g/mol
2-(Diphenylphosphino)ethylamine, is used as a fine chemical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Diphenylphosphino)ethylamine, is used as a fine chemical intermediate. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Air Sensitive.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4848-43-5
Molecular Formula C14H16NP
Molecular Weight (g/mol) 229.26
MDL Number MFCD00233831
InChI Key RXEPBCWNHKZECN-UHFFFAOYSA-N
Synonym 2-diphenylphosphino ethylamine, 2-diphenylphosphino ethanamine, 2-aminoethyl diphenylphosphine, 1-amino-2-diphenylphosphino ethane, 2-aminoethyl diphenylphosphane, 2-diphenylphosphanyl ethan-1-amine, 2-diphenylphosphino ethanamine hydrochloride, acmc-209v7l, nh2 ch2 2pph2, 2-diphenylphosphino-ethylamine
PubChem CID 273264
IUPAC Name 2-diphenylphosphanylethanamine
SMILES NCCP(C1=CC=CC=C1)C1=CC=CC=C1
Specifications
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 229.27
Percent Purity 95%
Chemical Name or Material 2-(Diphenylphosphino)ethylamine
compliance-icons
Product Identifier
  • 2-(Diphenylphosphino)ethylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 2-(Diphenylphosphino)ethylamine