accessibility menu, dialog, popup

UserName

CAS RN 4848-43-5

IUPAC Name: (2-aminoethyl)diphenylphosphane
Synonyms: (2-aminoethyl)diphenylphosphane
Molecular Weight (g/mol): 229.26
Molecular Formula: C14H16NP
InChi Key: RXEPBCWNHKZECN-UHFFFAOYSA-N
SMILES: NCCP(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results