missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropropanol, TRC

Product Code. 30367040
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367040 500 mg 500mg
30355035 100 mg 100mg
30361994 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30367040 Supplier Toronto Research Chemicals Supplier No. TRCC788970500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-89-7
Molecular Formula C3 H7 Cl O
Molecular Weight (g/mol) 94.54
Synonym 2-Chloro-1-propanol, 1-Hydroxy-2-chloropropane, 2-Chloropropanol, 2-Chloropropyl alcohol, NSC 122672, Propylene chlorohydrin, Zetag 7664-propylene chlorohydrin copolymer, β-Propylene chlorohydrin
IUPAC Name 2-chloropropan-1-ol
SMILES CC(Cl)CO

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C3H7ClO/c1-3(4)2-5/h3,5H,2H2,1H3
Formula Weight 94.0185
Percent Purity >95%
Chemical Name or Material 2-Chloro-1-propanol
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.