CAS RN 78-89-7
CAS RN 78-89-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Chloropropanol, TRC
CAS: 78-89-7 Molecular Formula: C3 H7 Cl O Molecular Weight (g/mol): 94.54 Synonym: 2-Chloro-1-propanol,1-Hydroxy-2-chloropropane,2-Chloropropanol,2-Chloropropyl alcohol,NSC 122672,Propylene chlorohydrin,Zetag 7664-propylene chlorohydrin copolymer,β-Propylene chlorohydrin IUPAC Name: 2-chloropropan-1-ol SMILES: CC(Cl)CO