Learn More
2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine Hydrochloride, TRC
Product Code. 30352628
Change view
Click to view available options
Quantity:
2.5 g
5 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30352628 5 g 5g
30365264 2.5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30352628 Supplier Toronto Research Chemicals Supplier No. TRCC3692405G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 86604-75-3
Molecular Formula C9 H12 Cl N O . Cl H
Molecular Weight (g/mol) 222.11
Synonym 2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride (1:1), Pyridine, 2-(chloromethyl)-4-methoxy-3,5-dimethyl-, hydrochloride (9CI), 2-(Chloromethyl)-3,5-dimethyl-4-methoxypyridine hydrochloride, 2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride, 3,5-Dimethyl-2-chloromethyl-4-methoxypyridine hydrochloride, Cc-993
IUPAC Name 2-(chloromethyl)-4-methoxy-3,5-dimethylpyridine;hydrochloride
SMILES Cl.COc1c(C)cnc(CCl)c1C
Spezifikation
Quantity 5 g
InChI Formula InChI=1S/C9H12ClNO.ClH/c1-6-5-11-8(4-10)7(2)9(6)12-3;/h5H,4H2,1-3H3;1H
Formula Weight 221.0374
Chemical Name or Material 2-Chloromethyl-3,5-dimethyl-4-methoxypyridine Hydrochloride