accessibility menu, dialog, popup

UserName

CAS RN 86604-75-3

IUPAC Name: 2-(chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride
Synonyms: 2-(chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride
Molecular Weight (g/mol): 222.11
Molecular Formula: C9H13Cl2NO
InChi Key: LCJDHJOUOJSJGS-UHFFFAOYSA-N
SMILES: Cl.COC1=C(C)C(CCl)=NC=C1C
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results