Learn More
2-Chloromalonaldehyde, TRC
Product Code. 30133995
Change view
Click to view available options
Quantity:
500 mg
2.5 g
5 g
Unit Size:
2.50g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133995 2.5 g 2.50g
30136221 5 g 5g
30004658 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133995 Supplier Toronto Research Chemicals Supplier No. TRCC3683202.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 36437-19-1
Molecular Formula C3 H3 Cl O2
Molecular Weight (g/mol) 106.508
Synonym 2-Chloro-1,3-propanedial, Chloromalonaldehyde, Chloromalondialdehyde, Chloromalonic Dialdehyde
IUPAC Name 2-chloropropanedial
SMILES ClC(C=O)C=O
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C3H3ClO2/c4-3(1-5)2-6/h1-3H
Formula Weight 105.982
Chemical Name or Material 2-Chloromalonaldehyde