accessibility menu, dialog, popup

UserName

CAS RN 36437-19-1

OHClO(Z)

CAS RN 36437-19-1

IUPAC Name: (2Z)-2-chloro-3-hydroxyprop-2-enal
Synonyms: (2Z)-2-chloro-3-hydroxyprop-2-enal
Molecular Weight (g/mol): 106.51
Molecular Formula: C3H3ClO2
InChi Key: SYXFXUMRQSTIKC-IWQZZHSRSA-N
SMILES: O\C=C(/Cl)C=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)