Learn More
2-Chlorobenzaldehyde, Sigma-Aldrich™
Product Code. 15860871
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15860871 1 L 1 L
15850871 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15860871 Supplier Sigma Aldrich Supplier No. 8202761000
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 89-98-5
Grade Research
Molecular Weight (g/mol) 140.57
Specifications
Melting Point 10°C
Density 1.25 g/cm3 at 20°C
MDL Number MFCD00003304
Quantity 1 L
Synonym 2-Chlorobenzaldehyde, 2-Chlorobenzaldehyde