accessibility menu, dialog, popup

UserName

ClO

CAS RN 89-98-5

IUPAC Name: 2-chlorobenzaldehyde
Synonyms: 2-chlorobenzaldehyde
Molecular Weight (g/mol): 140.57
Molecular Formula: C7H5ClO
InChi Key: FPYUJUBAXZAQNL-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
  • (7)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (5)
  • (3)
  • (3)
  • (3)
  • (4)
  • (7)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)
  • (3)
  • (3)
  • (5)
  • (5)
  • (2)