missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-(trifluoromethyl)phenyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 11327846
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327846 5 g 5g
11317846 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327846 Supplier Thermo Scientific Alfa Aesar Supplier No. L13016.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23165-49-3
Molecular Formula C8H3ClF3NS
Molecular Weight (g/mol) 237.624
MDL Number MFCD00041055
InChI Key KHTMKXDMVYHDSY-UHFFFAOYSA-N
PubChem CID 141005
IUPAC Name 1-chloro-2-isothiocyanato-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)N=C=S)Cl

Specifications

Density 1.45
Boiling Point 239°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.57
Quantity 5 g
UN Number UN2922
Beilstein 2112809
Sensitivity Moisture sensitive
Synonym 2-chloro-5-trifluoromethyl phenyl isothiocyanate, 1-chloro-2-isothiocyanato-4-trifluoromethyl benzene, 2-chloro-5-trifluoromethylphenyl isothiocyanate, 2-chloro-5-trifluoromethyl phenylisothiocyanate, benzene, 1-chloro-2-isothiocyanato-4-trifluoromethyl, benzene,1-chloro-2-isothiocyanato-4-trifluoromethyl, 2-chloro-5-trifluoromethyl benzenisothiocyanate, pubchem5015, acmc-1ciqp
Formula Weight 237.63
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)phenyl isothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.