accessibility menu, dialog, popup

UserName

FFFClNCS

CAS RN 23165-49-3

IUPAC Name: 1-chloro-2-isothiocyanato-4-(trifluoromethyl)benzene
Synonyms: 1-chloro-2-isothiocyanato-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 237.62
Molecular Formula: C8H3ClF3NS
InChi Key: KHTMKXDMVYHDSY-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(Cl)C(=C1)N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results