missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluoro-6-methylpyridine, 95%, Thermo Scientific™

Product Code. 15481509
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15481509 1 g 1g
15491509 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15481509 Supplier Thermo Scientific Alfa Aesar Supplier No. H64347.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolitik

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 884494-78-4
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.561
MDL Number MFCD04972391
InChI Key ICSRNKKGNHKSJH-UHFFFAOYSA-N
Synonym 2-chloro-5-fluoro-6-methylpyridine, 2-chloro-5-fluoro-6-picoline, 6-chloro-3-fluoro-2-methyl-pyridine, 6-chloro-3-fluoro-2-picoline, pubchem6219, ksc659q3d, pyridine,6-chloro-3-fluoro-2-methyl, 6-chloranyl-3-fluoranyl-2-methyl-pyridine
PubChem CID 19867376
IUPAC Name 6-chloro-3-fluoro-2-methylpyridine
SMILES CC1=C(C=CC(=N1)Cl)F

Tekniske data

Quantity 1 g
Beilstein 13638700
Formula Weight 145.56
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Chloro-5-fluoro-6-methylpyridine
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.