accessibility menu, dialog, popup

UserName

CAS RN 884494-78-4

CH3NClF

CAS RN 884494-78-4

IUPAC Name: 6-chloro-3-fluoro-2-methylpyridine
Synonyms: 6-chloro-3-fluoro-2-methylpyridine
Molecular Weight (g/mol): 145.56
Molecular Formula: C6H5ClFN
InChi Key: ICSRNKKGNHKSJH-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results