missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-nitro-5-(trifluoromethyl)pyridine, 95%

Product Code. 15411589
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15411589 1 g 1g
15421589 5 g 5g
15438886 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15411589 Supplier Thermo Scientific Alfa Aesar Supplier No. H64394.03
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 72587-15-6 | C6H2ClF3N2O2 | 226.539 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72587-15-6
Molecular Formula C6H2ClF3N2O2
Molecular Weight (g/mol) 226.539
MDL Number MFCD06255171
InChI Key DRPIKFKCAJGTJF-UHFFFAOYSA-N
Synonym 2-chloro-3-nitro-5-trifluoromethyl pyridine, 2-chloro-5-trifluoromethyl-3-nitropyridine, pyridine, 2-chloro-3-nitro-5-trifluoromethyl, 5-trifluoromethyl-3-nitro-2-chloropyridine, pubchem17529, acmc-209onn, abbypharma ap-11-5568, 2-chloro-3-nitro-5-trifluoromethyl-pyridine, 2-chloro3-nitro-5-trifluoromethyl pyridine
PubChem CID 4736950
IUPAC Name 2-chloro-3-nitro-5-(trifluoromethyl)pyridine
SMILES C1=C(C=NC(=C1[N+](=O)[O-])Cl)C(F)(F)F

Specifications

Color Colorless to Yellow
Quantity 1 g
Beilstein 7586778
Formula Weight 226.54
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Chloro-3-nitro-5-(trifluoromethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.