accessibility menu, dialog, popup

UserName

NNOOClFFF

CAS RN 72587-15-6

IUPAC Name: 2-chloro-3-nitro-5-(trifluoromethyl)pyridine
Synonyms: 2-chloro-3-nitro-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 226.54
Molecular Formula: C6H2ClF3N2O2
InChi Key: DRPIKFKCAJGTJF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Cl)N=CC(=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results