Learn More
2-Chloro-3-cyano-6-methylpyridine, 99%
Product Code. 11467557
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467557 1 g 1g
11477557 5 g 5g
11487557 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11467557 Supplier Thermo Scientific Alfa Aesar Supplier No. A15648.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 28900-10-9 | C7H5ClN2 | 152.58 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28900-10-9
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.58
MDL Number MFCD00082767
InChI Key YSBNBAYNISAUIT-UHFFFAOYSA-N
Synonym 2-chloro-6-methylnicotinonitrile, 2-chloro-3-cyano-6-methylpyridine, 2-chloro-6-methyl-3-pyridinecarbonitrile, 2-chloro-3-cyano-6-picoline, 3-pyridinecarbonitrile, 2-chloro-6-methyl, 2-chloro-6-methyl-nicotinonitrile, pubchem6128, 2-chloro-6-methylnicotinonitrile #, 2-chloro-3-cyano-6-methyl pyridine, 2-chloro-3-cyano-6-methyl-pyridine
PubChem CID 520400
IUPAC Name 2-chloro-6-methylpyridine-3-carbonitrile
SMILES CC1=CC=C(C#N)C(Cl)=N1
Specifications
Melting Point 114°C to 117°C
Quantity 1 g
Beilstein 120414
Formula Weight 152.58
Percent Purity 99%
Chemical Name or Material 2-Chloro-3-cyano-6-methylpyridine