accessibility menu, dialog, popup

UserName

CAS RN 28900-10-9

CH3NClN

CAS RN 28900-10-9

IUPAC Name: 2-chloro-6-methylpyridine-3-carbonitrile
Synonyms: 2-chloro-6-methylpyridine-3-carbonitrile
Molecular Weight (g/mol): 152.58
Molecular Formula: C7H5ClN2
InChi Key: YSBNBAYNISAUIT-UHFFFAOYSA-N
SMILES: CC1=CC=C(C#N)C(Cl)=N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)