missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-chlorotoluene, 99%, Thermo Scientific™

Product Code. 11441727
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441727 25 g 25g
11431727 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441727 Supplier Thermo Scientific Alfa Aesar Supplier No. B25697.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62356-27-8
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.48
MDL Number MFCD00061121
InChI Key DMARBQGIQKLIPM-UHFFFAOYSA-N
Synonym 2-bromo-6-chlorotoluene, benzene, 1-bromo-3-chloro-2-methyl, pubchem3601, 2-chloro-6-bromotoluene, 6-bromo-2-chlorotoluene, acmc-209n2m, ksc357m2t, attercop-chm at132933, #, 1-bromanyl-3-chloranyl-2-methyl-benzene
PubChem CID 123552
IUPAC Name 1-bromo-3-chloro-2-methylbenzene
SMILES CC1=C(Cl)C=CC=C1Br

Specifications

Density 1.58
Quantity 25 g
Refractive Index 1.578
Boiling Point 120°C (40 mmHg)
Formula Weight 205.48
Percent Purity 99%
Chemical Name or Material 2-Bromo-6-chlorotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.