accessibility menu, dialog, popup

UserName

CH3ClBr

CAS RN 62356-27-8

IUPAC Name: 1-bromo-3-chloro-2-methylbenzene
Synonyms: 1-bromo-3-chloro-2-methylbenzene
Molecular Weight (g/mol): 205.48
Molecular Formula: C7H6BrCl
InChi Key: DMARBQGIQKLIPM-UHFFFAOYSA-N
SMILES: CC1=C(Cl)C=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results