missing translation for 'onlineSavingsMsg'
Learn More

2'-Amino-4',5'-methylenedioxyacetophenone hydrochloride, 98%, Thermo Scientific™

Product Code. 11460437
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460437 100 g 100g
11440437 5 g 5g
11450437 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460437 Supplier Thermo Scientific Alfa Aesar Supplier No. B21788.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93983-01-8
Molecular Formula C9H10ClNO3
Molecular Weight (g/mol) 215.633
MDL Number MFCD00053022
InChI Key PIVDPBIZDWHINL-UHFFFAOYSA-N
Synonym 2'-amino-4',5'-methylenedioxyacetophenone hydrochloride, 1-6-aminobenzo d 1,3 dioxol-5-yl ethanone hydrochloride, 1-6-amino-1,3-benzodioxol-5-yl ethan-1-one hydrochloride, 2-amino-4,5-methylenedioxyacetophenone hydrochloride, 1-6-amino-2h-1,3-benzodioxol-5-yl ethanone hydrochloride, 1-6-amino-1,3-benzodioxol-5-yl ethanone hydrochloride, 1-6-azanyl-1,3-benzodioxol-5-yl ethanone hydrochloride
PubChem CID 16205927
IUPAC Name 1-(6-amino-1,3-benzodioxol-5-yl)ethanone;hydrochloride
SMILES CC(=O)C1=CC2=C(C=C1N)OCO2.Cl

Specifications

Quantity 100 g
Sensitivity Hygroscopic
Formula Weight 215.64
Percent Purity 98%
Chemical Name or Material 2'-Amino-4',5'-methylenedioxyacetophenone hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.