accessibility menu, dialog, popup

UserName

CAS RN 93983-01-8

IUPAC Name: hydrogen 1-(6-amino-2H-1,3-benzodioxol-5-yl)ethan-1-one chloride
Synonyms: hydrogen 1-(6-amino-2H-1,3-benzodioxol-5-yl)ethanone chloride
Molecular Weight (g/mol): 215.63
Molecular Formula: C9H10ClNO3
InChi Key: PIVDPBIZDWHINL-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC(=O)C1=CC2=C(OCO2)C=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results