missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylphenylpropionate, TRC

Product Code. 30374881
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374881 1 g
30371716 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30374881 Supplier Toronto Research Chemicals Supplier No. TRCD4770201G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51233-80-8
Molecular Formula C11 H14 O2
Molecular Weight (g/mol) 178.23
Synonym Phenol, 2,6-dimethyl-, 1-propanoate, Phenol, 2,6-dimethyl-, propanoate (9CI), 2,6-Dimethylphenyl propanoate, 2,6-Dimethylphenyl propionate, 2,6-Xylyl propionate, Propanoic acid 2,6-dimethylphenyl ester
IUPAC Name 3-(2,6-dimethylphenyl)propanoic acid
SMILES Cc1cccc(C)c1CCC(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C11H14O2/c1-8-4-3-5-9(2)10(8)6-7-11(12)13/h3-5H,6-7H2,1-2H3,(H,12,13)
Formula Weight 178.0994
Chemical Name or Material 2,6-Dimethylphenylpropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.