CAS RN 51233-80-8
CAS RN 51233-80-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,6-Dimethylphenylpropionate, TRC
CAS: 51233-80-8 Molecular Formula: C11 H14 O2 Molecular Weight (g/mol): 178.23 Synonym: Phenol, 2,6-dimethyl-, 1-propanoate,Phenol, 2,6-dimethyl-, propanoate (9CI),2,6-Dimethylphenyl propanoate,2,6-Dimethylphenyl propionate,2,6-Xylyl propionate,Propanoic acid 2,6-dimethylphenyl ester IUPAC Name: 3-(2,6-dimethylphenyl)propanoic acid SMILES: Cc1cccc(C)c1CCC(=O)O