missing translation for 'onlineSavingsMsg'
Learn More

2,4-Quinolinediol, 97%, Thermo Scientific™

Product Code. p-2697122 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10468290 10 g Glass bottle 10g
10326780 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10468290 Supplier Thermo Scientific Acros Supplier No. 132200100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-95-3
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
InChI Key HDHQZCHIXUUSMK-UHFFFAOYSA-N
Synonym 2,4-quinolinediol, 2,4-dihydroxyquinoline, 4-hydroxyquinolin-2 1h-one, quinoline-2,4-diol, 4-hydroxy-2-quinolone, 4-hydroxycarbostyril, 2 1h-quinolinone, 4-hydroxy, 4-hydroxy-2-quinolinone, hydroxycarbostyril, 2-hydroxyquinolin-4 1h-one
PubChem CID 54680871
ChEBI CHEBI:75926
IUPAC Name 4-hydroxy-1H-quinolin-2-one
SMILES C1=CC=C2C(=C1)C(=CC(=O)N2)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 10 g
Beilstein 21, 171
Formula Weight 161.16
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 2, 4-Quinolinediol
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Quinolinediol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.