accessibility menu, dialog, popup

UserName

HOONH

CAS RN 86-95-3

IUPAC Name: 2-hydroxy-1,4-dihydroquinolin-4-one
Synonyms: 2-hydroxy-1H-quinolin-4-one
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: HDHQZCHIXUUSMK-UHFFFAOYSA-N
SMILES: OC1=CC(=O)C2=CC=CC=C2N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results