Learn More
2,4-Dimethyl-1-heptene, TRC
Product Code. 30383573
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383573 100 mg
30376115 50 mg
30376221 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30383573 Supplier Toronto Research Chemicals Supplier No. TRCD460298100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 19549-87-2
Molecular Formula C9H18
Molecular Weight (g/mol) 126.23
Synonym 2,4-Dimethyl-1-heptene
SMILES CCCC(C)CC(=C)C
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C9H18/c1-5-6-9(4)7-8(2)3/h9H,2,5-7H2,1,3-4H3
Formula Weight 126.1409
Chemical Name or Material 2,4-Dimethyl-1-heptene