CAS RN 19549-87-2
CAS RN 19549-87-2
IUPAC Name:
2,4-dimethylhept-1-ene
Synonyms:
2,4-dimethylhept-1-ene
Molecular Weight (g/mol):
126.24
Molecular Formula:
C9H18
InChi Key:
CZGAOHSMVSIJJZ-UHFFFAOYNA-N
SMILES:
CCCC(C)CC(C)=C
1
–
1
of
1
results
1
2,4-Dimethyl-1-heptene, TRC
CAS: 19549-87-2 Molecular Formula: C9H18 Molecular Weight (g/mol): 126.23 Synonym: 2,4-Dimethyl-1-heptene SMILES: CCCC(C)CC(=C)C