Learn More
2,4-Difluorobenzyl bromide, 98+%
Product Code. 11432238
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432238 25 g 25g
11422238 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11432238 Supplier Thermo Scientific Alfa Aesar Supplier No. A15213.14
Request Bulk or Custom Format
CAS: 23915-07-3 | C7H5BrF2 | 207.018 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 23915-07-3
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.018
MDL Number MFCD00011649
InChI Key IBLMYGXJKQIGSN-UHFFFAOYSA-N
Synonym 2,4-difluorobenzyl bromide, 1-bromomethyl-2,4-difluorobenzene, 2,4-difluorobenzylbromide, alpha-bromo-2,4-difluorotoluene, benzene, 1-bromomethyl-2,4-difluoro, 2,4-difluoro benzyl bromide, 1-bromomethyl-2,4-difluoro-benzene, 2-chloromethyl quinoline hcl salt, fr cf d1e, pubchem4911
PubChem CID 90297
IUPAC Name 1-(bromomethyl)-2,4-difluorobenzene
SMILES C1=CC(=C(C=C1F)F)CBr
Specifications
Density 1.613
Boiling Point 42°C to 43°C (1 mmHg)
Flash Point 85°C (185°F)
Refractive Index 1.525
Quantity 25 g
UN Number UN1760
Beilstein 4177539
Formula Weight 207.02
Percent Purity ≥98%
Chemical Name or Material 2,4-Difluorobenzyl bromide