CAS RN 23915-07-3
CAS RN 23915-07-3
IUPAC Name:
1-(bromomethyl)-2,4-difluorobenzene
Synonyms:
1-(bromomethyl)-2,4-difluorobenzene
Molecular Weight (g/mol):
207.02
Molecular Formula:
C7H5BrF2
InChi Key:
IBLMYGXJKQIGSN-UHFFFAOYSA-N
SMILES:
FC1=CC=C(CBr)C(F)=C1
1
–
2
of
2
results
1
2,4-Difluorobenzyl bromide, 98+%
CAS: 23915-07-3 Molecular Formula: C7H5BrF2 Molecular Weight (g/mol): 207.018 MDL Number: MFCD00011649 InChI Key: IBLMYGXJKQIGSN-UHFFFAOYSA-N Synonym: 2,4-difluorobenzyl bromide,1-bromomethyl-2,4-difluorobenzene,2,4-difluorobenzylbromide,alpha-bromo-2,4-difluorotoluene,benzene, 1-bromomethyl-2,4-difluoro,2,4-difluoro benzyl bromide,1-bromomethyl-2,4-difluoro-benzene,2-chloromethyl quinoline hcl salt,fr cf d1e,pubchem4911 PubChem CID: 90297 IUPAC Name: 1-(bromomethyl)-2,4-difluorobenzene SMILES: C1=CC(=C(C=C1F)F)CBr