missing translation for 'onlineSavingsMsg'
Learn More

2,4'-Dibromobenzophenone, TRC

Product Code. 30365979
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365979 500 mg 500mg
30358296 100 mg 100mg
30352791 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30365979 Supplier Toronto Research Chemicals Supplier No. TRCD082320500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78281-59-1
Molecular Formula C13 H8 Br2 O
Molecular Weight (g/mol) 340.01
Synonym (2-Bromophenyl)(4-bromophenyl)methanone, 2,4'-Dibromobenzophenone
IUPAC Name (2-bromophenyl)-(4-bromophenyl)methanone
SMILES Brc1ccc(cc1)C(=O)c2ccccc2Br

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C13H8Br2O/c14-10-7-5-9(6-8-10)13(16)11-3-1-2-4-12(11)15/h1-8H
Formula Weight 337.8942
Chemical Name or Material 2,4'-Dibromobenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.